Learn More
2,4,6-Trichloropyrimidine, 98%, Thermo Scientific Chemicals
Product Code. 11425243
Click to view available options
Quantity:
5 g
25 g
100 g
It is applied In selective Suzuki coupling with arylboronic acids has been reported to give 6-arylpyrimidines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied In selective Suzuki coupling with arylboronic acids has been reported to give 6-arylpyrimidines.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3764-01-0
Molecular Formula C4HCl3N2
Molecular Weight (g/mol) 183.416
MDL Number MFCD00006063
InChI Key DPVIABCMTHHTGB-UHFFFAOYSA-N
Synonym pyrimidine, 2,4,6-trichloro, 2,4,6-trichloro pyrimidine, 2,4,6-trichloro-pyrimidine, ccris 7597, 2,4,6-trichlorpyrimidin, pubchem9310, acmc-1agpv, 2,6-trichloropyrimidine, 2.4.6-trichloropyrimidin, dsstox_cid_29143
PubChem CID 77378
IUPAC Name 2,4,6-trichloropyrimidine
SMILES C1=C(N=C(N=C1Cl)Cl)Cl
Specifications
Melting Point 21°C to 25°C
Density 1.595
Boiling Point 213°C to 215°C
Flash Point >110°C (230°F)
Odor Irritating
Refractive Index 1.57
Quantity 5 g
UN Number UN1759
Beilstein 118284
Solubility Information Insoluble in water.
Formula Weight 183.43
Percent Purity 98%
Chemical Name or Material 2,4,6-Trichloropyrimidine