missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetramethyl-2-cyclopentenone, cis + trans, 95%

Product Code. p-7045153
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 54458-61-6 | C9H14O | 138.21 g/mol

2,3,4,5-Tetramethyl-2-cyclopentenone was used in the synthesis of chiral pre-ligands, (R)-3,3?-bis(tetramethylcyclopentadienyl)-2,2?-bismethoxy-1,1?-bisnaphthalene and (R)-3-tetramethylcyclopentadienyl-2,2?-bismethoxy-1,1?-bisnaphthalene by reacting with (R)-3,3?-dilithium-2,2?-bismethoxy-1,1?-bisnaphthalene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4,5-Tetramethyl-2-cyclopentenone was used in the synthesis of chiral pre-ligands, (R)-3,3′-bis(tetramethylcyclopentadienyl)-2,2′-bismethoxy-1,1′-bisnaphthalene and (R)-3-tetramethylcyclopentadienyl-2,2′-bismethoxy-1,1′-bisnaphthalene by reacting with (R)-3,3′-dilithium-2,2′-bismethoxy-1,1′-bisnaphthalene.

Solubility
Not miscible with water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54458-61-6
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
MDL Number MFCD00010248
InChI Key ARUAYSANQMCCEN-UHFFFAOYSA-N
Synonym 2,3,4,5-tetramethylcyclopent-2-enone, 2,3,4,5-tetramethyl-2-cyclopentenone, 2,3,4,5-tetramethyl-2-cyclopenten-1-one, 2-cyclopenten-1-one, 2,3,4,5-tetramethyl, 2,3,4,5-tetramethyl-2-cyclopentenone, cis + trans, tetramethylcyclopent-2-enone, pubchem15060, acmc-1asp4, 2,3,4,5-tetramethylcyclopentenone, 2,3,4,5-tetramethyl-cyclopent-2-enone
PubChem CID 41092
IUPAC Name 2,3,4,5-tetramethylcyclopent-2-en-1-one
SMILES CC1C(C(=O)C(=C1C)C)C

Specifications

Density 0.917
Boiling Point 99°C to 100°C (30 mmHg)
Flash Point 73°C (163°F)
Refractive Index 1.476
Quantity 5 g
Beilstein 2324088
Solubility Information Not miscible with water.
Formula Weight 138.21
Percent Purity 95%
Chemical Name or Material 2,3,4,5-Tetramethyl-2-cyclopentenone, Cis + Trans

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis