Learn More
2,2,2-Trifluoroethanol, ≥99%, Reagent Grade, Honeywell™
Product Code. p-7120042 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
Reagent Grade,≥99%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 75-89-8
Molecular Formula C2H3F3O
Molecular Weight (g/mol) 100.04
MDL Number MFCD00004672
InChI Key RHQDFWAXVIIEBN-UHFFFAOYSA-N
Synonym trifluoroethanol, ethanol, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-ol, fluorinol 85, 2,2,2-trifluoroethyl alcohol, trifluoroethyl alcohol, trifluoro ethanol, tfe, perfluoro-1,1-dihydroethanol, tfetoh
PubChem CID 6409
ChEBI CHEBI:42330
SMILES OCC(F)(F)F
Specifications
Melting Point -44°C
Color Colorless
Density 1.373g/mL at 25°C
Boiling Point 73°C to 77°C
Flash Point 29°C (84.2°F)
Odor Characteristic
Packaging Glass bottle
Linear Formula CF3CH2OH
Quantity 25 g
UN Number UN1986
Beilstein 1733203
IUPAC Name 2,2,2-trifluoroethan-1-ol
Vapor Density 3.5 (vs air)
Vapor Pressure 70mmHg (25°C)
Percent Purity ≥99%
Chemical Name or Material 2,2,2-Trifluoroethanol