Learn More
1-Octanol, ReagentPlus™, 99%, Honeywell™
Product Code. 15621040 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Reagent Grade, 99%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 111-87-5
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00002988
InChI Key KBPLFHHGFOOTCA-UHFFFAOYSA-N
Synonym 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
PubChem CID 957
ChEBI CHEBI:16188
IUPAC Name octan-1-ol
SMILES CCCCCCCCO
Specifications
Melting Point -15°C
Color Colorless
Density 0.827g/mL at 25°C
Boiling Point 196°C
Flash Point 80°C (176°F)
Packaging Stainless Steel Can
Linear Formula CH3(CH2)7OH
Refractive Index n20/D 1.429(lit.)
Quantity 20 L
Beilstein 1697461
Vapor Density 4.5 (vs air)
Formula Weight 130.23g/mol
Percent Purity 99%
Grade ReagentPlus™
Physical Form Liquid
Chemical Name or Material 1-Octanol