missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-methoxynaphthalene, 97%, Thermo Scientific Chemicals

Product Code. 11403178
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 3401-47-6 | C11H9BrO | 237.10 g/mol

1-Bromo-2-methoxy-naphthalene is a useful synthetic intermediate. It is used as a reagent to synthesize a catalyst for highly enantioselective aziridination of styrene derivatives. It can also be used to prepare biaryls or biheterocycles by palladium-catalyzed Ullmann coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-methoxy-naphthalene is a useful synthetic intermediate. It is used as a reagent to synthesize a catalyst for highly enantioselective aziridination of styrene derivatives. It can also be used to prepare biaryls or biheterocycles by palladium-catalyzed Ullmann coupling.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3401-47-6
Molecular Formula C11H9BrO
Molecular Weight (g/mol) 237.10
MDL Number MFCD00055374
InChI Key XNIGURFWNPLWJM-UHFFFAOYSA-N
Synonym 1-bromo-2-methoxy-naphthalene, naphthalene, 1-bromo-2-methoxy, acmc-1cjky, maybridge3_005134, 2-methoxy-1-bromonaphthalene, 1-bromo-2-methoxy-naphtalene
PubChem CID 72860
IUPAC Name 1-bromo-2-methoxynaphthalene
SMILES COC1=CC=C2C=CC=CC2=C1Br

Specifications

Melting Point 79°C to 80°C
Quantity 25 g
Beilstein 2045600
Solubility Information Slightly soluble in water.
Formula Weight 237.1
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-methoxynaphthalene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.